missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

alpha-Ketoglutaric acid disodium salt dihydrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J6315403

 View more versions of this product

Catalog No. AAJ6315403



Description

Description

alpha-Ketoglutaric acid disodium salt dihydrate is used as pharmaceutical intermediate. α-Ketoglutaric acid disodium salt dihydrate is suitable reagent used in a study to investigate the role of nitric oxide (NO) in mast cell physiology and biochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
alpha-Ketoglutaric acid disodium salt dihydrate is used as pharmaceutical intermediate. α-Ketoglutaric acid disodium salt dihydrate is suitable reagent used in a study to investigate the role of nitric oxide (NO) in mast cell physiology and biochemistry.

Solubility
Soluble in water (1 M).

Notes
Hygroscopic. Store under inert gas. Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

alpha-Ketoglutaric acid disodium salt dihydrate
White
MFCD00150702
Hygroscopic
Soluble in water (1 M).
C(CC(=O)[O-])C(=O)C(=O)[O-].[Na+].[Na+]
190.062
226.09 (190.06 Anhydrous)
Powder
305-72-6
C5H4Na2O5
1 g
disodium 2-oxoglutarate, unii-flp7p4rm46, disodium 2-oxopentanedioate, flp7p4rm46, pentanedioic acid, 2-oxo-, disodium salt, 2-oxoglutaric acid disodium salt, 2-ketoglutaric acid disodium salt, alpha-ketoglutaric acid sodium salt, disodium ? ketoglutarate, disodium oxoglurate
YBGBJYVHJTVUSL-UHFFFAOYSA-L
disodium;2-oxopentanedioate
31040
99%

Chemical Identifiers

305-72-6
190.062
YBGBJYVHJTVUSL-UHFFFAOYSA-L
31040
C(CC(=O)[O-])C(=O)C(=O)[O-].[Na+].[Na+]
C5H4Na2O5
MFCD00150702
disodium 2-oxoglutarate, unii-flp7p4rm46, disodium 2-oxopentanedioate, flp7p4rm46, pentanedioic acid, 2-oxo-, disodium salt, 2-oxoglutaric acid disodium salt, 2-ketoglutaric acid disodium salt, alpha-ketoglutaric acid sodium salt, disodium ? ketoglutarate, disodium oxoglurate
disodium;2-oxopentanedioate
Safety and Handling

Safety and Handling

EINECSNumber : 206-167-8

RTECSNumber : SA0454700

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only