Learn More
alpha-Ketoglutaric acid disodium salt dihydrate, 99%
CAS: 305-72-6 | C5H4Na2O5 | 190.062 g/mol
Supplier: Thermo Scientific Chemicals J6315430
Description
alpha-Ketoglutaric acid disodium salt dihydrate is used as pharmaceutical intermediate. α-Ketoglutaric acid disodium salt dihydrate is suitable reagent used in a study to investigate the role of nitric oxide (NO) in mast cell physiology and biochemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsalpha-Ketoglutaric acid disodium salt dihydrate is used as pharmaceutical intermediate. α-Ketoglutaric acid disodium salt dihydrate is suitable reagent used in a study to investigate the role of nitric oxide (NO) in mast cell physiology and biochemistry.
Solubility
Soluble in water (1 M).
Notes
Hygroscopic. Store under inert gas. Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
Specifications
| alpha-Ketoglutaric acid disodium salt dihydrate | |
| White | |
| MFCD00150702 | |
| Hygroscopic | |
| Soluble in water (1 M). | |
| C(CC(=O)[O-])C(=O)C(=O)[O-].[Na+].[Na+] | |
| 190.062 | |
| 226.09 (190.06 Anhydrous) | |
| Powder |
| 305-72-6 | |
| C5H4Na2O5 | |
| 250 g | |
| disodium 2-oxoglutarate, unii-flp7p4rm46, disodium 2-oxopentanedioate, flp7p4rm46, pentanedioic acid, 2-oxo-, disodium salt, 2-oxoglutaric acid disodium salt, 2-ketoglutaric acid disodium salt, alpha-ketoglutaric acid sodium salt, disodium ? ketoglutarate, disodium oxoglurate | |
| YBGBJYVHJTVUSL-UHFFFAOYSA-L | |
| disodium;2-oxopentanedioate | |
| 31040 | |
| 99% |
Chemical Identifiers
| 305-72-6 | |
| 190.062 | |
| YBGBJYVHJTVUSL-UHFFFAOYSA-L | |
| 31040 | |
| C(CC(=O)[O-])C(=O)C(=O)[O-].[Na+].[Na+] |
| C5H4Na2O5 | |
| MFCD00150702 | |
| disodium 2-oxoglutarate, unii-flp7p4rm46, disodium 2-oxopentanedioate, flp7p4rm46, pentanedioic acid, 2-oxo-, disodium salt, 2-oxoglutaric acid disodium salt, 2-ketoglutaric acid disodium salt, alpha-ketoglutaric acid sodium salt, disodium ? ketoglutarate, disodium oxoglurate | |
| disodium;2-oxopentanedioate |
Safety and Handling
EINECSNumber : 206-167-8
RTECSNumber : SA0454700
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only