Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Hydroxyfarnesylphosphonic acid, 96%, in 500mul ethanol


Quantity 1mg

Manufacturer: alfa aesar™  J67097MCR

 View more versions of this product

Catalog No. AAJ67097MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 302.35
Chemical Name or Material alpha-Hydroxyfarnesylphosphonic acid
CAS 140633-12-1
Physical Form Liquid
Assay 96%
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H371
Highly flammable liquid and vapor.
May cause damage to organs.
Health Hazard 3 GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 -30° to -10°C
Name Note 1mg in 500μL ethanol
MDL Number MFCD00216148
Quantity 1mg
UN Number UN1170
DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: ETHANOL SOLUTIONS

Specifications

Chemical Name or Material alpha-Hydroxyfarnesylphosphonic acid
CAS 140633-12-1
Name Note 1mg in 500μL ethanol
Formula Weight 302.35
MDL Number MFCD00216148
UN Number UN1170
Accessories

Danger

GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H371
Highly flammable liquid and vapor.
May cause damage to organs.

-30° to -10°C

SDS & Safety Documents