Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Hydroxy-alpha-methylbenzyl phenyl ketone, 98%

$403.81 - $403.81

Chemical Identifiers

Chemical Name or Material alpha-Hydroxy-alpha-methylbenzyl phenyl ketone
CAS 5623-26-7
Molecular Formula C15H14O2
Formula Weight 226.28
MDL Number MFCD00051804
Beilstein 2049961
Synonym alpha-Methylbenzoin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0724018

  SDS
alfa aesar™
L0724018
50g Each for $403.81
Description & Specifications

Specifications

Formula Weight 226.28
Chemical Name or Material alpha-Hydroxy-alpha-methylbenzyl phenyl ketone
Assay 98%
Molecular Formula C15H14O2
MDL Number MFCD00051804
CAS 5623-26-7
Synonym alpha-Methylbenzoin
Melting Point 63° to 67°C
Beilstein 2049961
Quantity 50g