Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Gliadin (43-49)

For cellular and molecular biology applications

Specifications

Product Type alpha-Gliadin (43-49)
Molecular Weight 875.40
Storage Requirements -30° to -10°C
Sequence Tyr-Pro-Gln-Pro-Gln-Pro-Phe
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66240LB0

 
alfa aesar™
J66240LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66240MB

 
alfa aesar™
J66240MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type alpha-Gliadin (43-49)
Molecular Weight 875.40
Storage Requirements -30° to -10°C
Sequence Tyr-Pro-Gln-Pro-Gln-Pro-Phe
Format Solid
Common Name α-Gliadorphin (43-49)
Regulatory Status RUO
Purification Method HPLC