missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
alpha-Ethyl-3-nitrocinnamic acid, 99%, Thermo Scientific™
$73.92 - $73.92
Chemical Identifiers
CAS | 124525-55-9 |
---|---|
Molecular Formula | C11H11NO4 |
Molecular Weight (g/mol) | 221.212 |
MDL Number | MFCD00134948 |
InChI Key | QMUHBYNLXQHZNR-RMKNXTFCSA-N |
Synonym | e-2-3-nitrobenzylidene butanoic acid, alpha-ethyl-3-nitrocinnamic acid, 2e-2-3-nitrophenyl methylidene butanoic acid, 2-3-nitrobenzylidene butanoic acid, .alpha.-ethyl-3-nitrocinnamic acid, ^a-ethyl-3-nitrocinnamic acid, cinnamic acid, .alpha.-ethyl-m-nitro, butanoic acid, 2-3-nitrophenyl methylene, 2e-2-ethyl-3-3-nitrophenyl prop-2-enoic acid |
PubChem CID | 6059692 |
IUPAC Name | (2E)-2-[(3-nitrophenyl)methylidene]butanoic acid |
SMILES | CCC(=CC1=CC(=CC=C1)[N+](=O)[O-])C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0822106
|
Thermo Scientific Chemicals
L0822106 |
5 g |
Each for $73.92
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
124525-55-9 | |
221.212 | |
QMUHBYNLXQHZNR-RMKNXTFCSA-N | |
6059692 | |
CCC(=CC1=CC(=CC=C1)[N+](=O)[O-])C(=O)O |
C11H11NO4 | |
MFCD00134948 | |
e-2-3-nitrobenzylidene butanoic acid, alpha-ethyl-3-nitrocinnamic acid, 2e-2-3-nitrophenyl methylidene butanoic acid, 2-3-nitrobenzylidene butanoic acid, .alpha.-ethyl-3-nitrocinnamic acid, ^a-ethyl-3-nitrocinnamic acid, cinnamic acid, .alpha.-ethyl-m-nitro, butanoic acid, 2-3-nitrophenyl methylene, 2e-2-ethyl-3-3-nitrophenyl prop-2-enoic acid | |
(2E)-2-[(3-nitrophenyl)methylidene]butanoic acid |
Specifications
124525-55-9 | |
C11H11NO4 | |
5 g | |
e-2-3-nitrobenzylidene butanoic acid, alpha-ethyl-3-nitrocinnamic acid, 2e-2-3-nitrophenyl methylidene butanoic acid, 2-3-nitrobenzylidene butanoic acid, .alpha.-ethyl-3-nitrocinnamic acid, ^a-ethyl-3-nitrocinnamic acid, cinnamic acid, .alpha.-ethyl-m-nitro, butanoic acid, 2-3-nitrophenyl methylene, 2e-2-ethyl-3-3-nitrophenyl prop-2-enoic acid | |
CCC(=CC1=CC(=CC=C1)[N+](=O)[O-])C(=O)O | |
221.212 | |
221.21 | |
alpha-Ethyl-3-nitrocinnamic acid |
138°C to 142°C | |
MFCD00134948 | |
2618101 | |
QMUHBYNLXQHZNR-RMKNXTFCSA-N | |
(2E)-2-[(3-nitrophenyl)methylidene]butanoic acid | |
6059692 | |
99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures