missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-D-(+)-Melibiose

$309.96 - $309.96

Chemical Identifiers

CAS 585-99-9
Molecular Formula C12H22O11
MDL Number MFCD00198188
Synonym 6-O-alpha-D-Galactopyranosyl-D-glucose
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6043906 View Documents Alfa Aesar™
J6043906
5g Each for $309.96
 
Specifications

Chemical Identifiers

585-99-9
C12H22O11
MFCD00198188
6-O-alpha-D-Galactopyranosyl-D-glucose

Specifications

White
585-99-9
MFCD00198188
6-O-alpha-D-Galactopyranosyl-D-glucose
342.3
alpha-D-(+)-Melibiose
5g
C12H22O11
14,5820
Soluble in water at 100mg/ml
Crystalline Powder
Safety and Handling

Safety and Handling

TSCA : TSCA