missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-D-Lactose monohydrate

Manufacturer:  Alfa Aesar™ A110740B

 View more versions of this product

Catalog No. AAA110740B



Description

Description

Solubility
Soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications

Specifications

alpha-D-Lactose monohydrate
+52° (c=4.5 in water, 22h)
1000g
C12H24O12
3768231
alpha-d-lactose monohydrate, alpha-lactose monohydrate, lactose monohydrate, lactose, monohydrate, unii-ewq57q8i5x, respitose, alpha-lactose hydrate, a-lactose monohydrate, lactose monohydrate nf, ewq57q8i5x
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O
360.312
360.32 (342.30 Anhydrous)
Odorless
207°C to 211°C (decomposition)
5989-81-1
MFCD00150747
14,5343
WSVLPVUVIUVCRA-KPKNDVKVSA-N
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
104938

Chemical Identifiers

5989-81-1
360.312
WSVLPVUVIUVCRA-KPKNDVKVSA-N
104938
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O
C12H24O12
MFCD00150747
alpha-d-lactose monohydrate, alpha-lactose monohydrate, lactose monohydrate, lactose, monohydrate, unii-ewq57q8i5x, respitose, alpha-lactose hydrate, a-lactose monohydrate, lactose monohydrate nf, ewq57q8i5x
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
Safety and Handling

Safety and Handling

RTECSNumber : OD9625000

TSCA : TSCA

SDS Documents