missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-D-Glucose-1-phosphate disodium salt tetrahydrate, 98%

Manufacturer:  Alfa Aesar™ A1206406

 View more versions of this product

Catalog No. AAA1206406


 

Description

Description

Applications
alpha-D-Glucose-1-phosphate disodium salt tetra hydrate is a glucose molecule that can be used in cellular catabolism. It is used as an intermediate in chemical and pharmaceutical research.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
Specifications

Specifications

alpha-D-Glucose-1-phosphate disodium salt tetrahydrate
98%
MFCD00150839
C(C1C(C(C(C(O1)OP(=O)(O)O)O)O)O)O.[Na].[Na]
306.114
376.16 (304.10 Anhydrous)
5g
56401-20-8
C6H13Na2O9P
AQMGMJDBUKKFOG-PKXGBZFFSA-N
sodium;[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate
131855210
~138°C

Chemical Identifiers

56401-20-8
306.114
AQMGMJDBUKKFOG-PKXGBZFFSA-N
C6H13Na2O9P
MFCD00150839
131855210
Safety and Handling

Safety and Handling

Recommended Storage : −30°C to −10°C

SDS Documents