Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Cyclopentylmandelic acid, 97%

Chemical Identifiers

Chemical Name or Material alpha-Cyclopentylmandelic acid
CAS 427-49-6
Formula Weight 220.26
MDL Number MFCD00019296
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6623503

  SDS
alfa aesar™
H6623503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6623506

  SDS
alfa aesar™
H6623506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6623514

  SDS
alfa aesar™
H6623514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 145° to 151°C
Formula Weight 220.26
CAS 427-49-6
MDL Number MFCD00019296
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Chemical Name or Material alpha-Cyclopentylmandelic acid
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Solid
Boiling Point 100°C (0.5mmHg)
Quantity 1g
Color White to pale brown
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.