missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Cyclohexylmandelic acid, 97%

$96.38 - $368.57

Chemical Identifiers

CAS 4335-77-7
Molecular Formula C14H18O3
Molecular Weight (g/mol) 234.295
MDL Number MFCD00194543
InChI Key YTRNSQPXEDGWMR-UHFFFAOYSA-N
Synonym alpha-cyclohexylmandelic acid, 2-cyclohexylmandelic acid, cyclohexylphenylglycolic acid, lespedamine, hexahydrobenzilic acid, cyclohexylmandelic acid, cyclohexyl-hydroxy-phenyl-acetic acid, cyclohexyl hydroxy phenylacetic acid, phenyl-cyclohexylglycolic acid, rcc 32
PubChem CID 97700
IUPAC Name 2-cyclohexyl-2-hydroxy-2-phenylacetic acid
SMILES C1CCC(CC1)C(C2=CC=CC=C2)(C(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6623806 View Documents Alfa Aesar™
H6623806
5g Each for $96.38
 
AAH6623814 View Documents Alfa Aesar™
H6623814
25g Each for $368.57
 
Specifications

Chemical Identifiers

4335-77-7
234.295
YTRNSQPXEDGWMR-UHFFFAOYSA-N
97700
C1CCC(CC1)C(C2=CC=CC=C2)(C(=O)O)O
C14H18O3
MFCD00194543
alpha-cyclohexylmandelic acid, 2-cyclohexylmandelic acid, cyclohexylphenylglycolic acid, lespedamine, hexahydrobenzilic acid, cyclohexylmandelic acid, cyclohexyl-hydroxy-phenyl-acetic acid, cyclohexyl hydroxy phenylacetic acid, phenyl-cyclohexylglycolic acid, rcc 32
2-cyclohexyl-2-hydroxy-2-phenylacetic acid

Specifications

135°C (3mmHg)
170°C to 175°C
4335-77-7
MFCD00194543
YTRNSQPXEDGWMR-UHFFFAOYSA-N
2-cyclohexyl-2-hydroxy-2-phenylacetic acid
97700
Solid
alpha-Cyclohexylmandelic acid
Brown to White
5g
C14H18O3
alpha-cyclohexylmandelic acid, 2-cyclohexylmandelic acid, cyclohexylphenylglycolic acid, lespedamine, hexahydrobenzilic acid, cyclohexylmandelic acid, cyclohexyl-hydroxy-phenyl-acetic acid, cyclohexyl hydroxy phenylacetic acid, phenyl-cyclohexylglycolic acid, rcc 32
C1CCC(CC1)C(C2=CC=CC=C2)(C(=O)O)O
234.295
234.29
97%
Safety and Handling

Safety and Handling

RTECSNumber : GV2770000