Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Cyclodextrin hydrate

$76.39 - $934.19

Chemical Identifiers

Chemical Name or Material alpha-Cyclodextrin hydrate
CAS 51211-51-9
Molecular Formula C36H60O30·xH2O
Formula Weight 972.86 (Anhydrous)
MDL Number MFCD00150804
Beilstein 4227442
Merck Index 14,2718
Synonym Schardinger alpha-dextrin; Cyclohexaamylose
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1809206

  SDS
alfa aesar™
A1809206
5g Each for $76.39

AAA1809222

  SDS
alfa aesar™
A1809222
100g Each for $934.19
Description & Specifications

Specifications

Formula Weight 972.86 (Anhydrous)
Synonym Schardinger alpha-dextrin; Cyclohexaamylose
RTECS Number GU2292000
TSCA TSCA
Melting Point 279°C (dec.)
Chemical Name or Material alpha-Cyclodextrin hydrate
Optical Rotation +136° (c=10 in Water)
Merck Index 14,2718
Quantity 5g
CAS 51211-51-9
MDL Number MFCD00150804
Beilstein 4227442
Molecular Formula C36H60O30·xH2O

Applications
alpha-Cyclodextrin hydrate is used in the food industry. The structure of cyclodextrins allows them to form inclusion complexes with a wide variety of molecules. It lead to enzyme-like specificity in reactions.

Solubility
Soluble in water. 145 g/L at 25

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store in the dark.