Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Cyano-4-methoxycinnamic acid, 98+%

$105.58 - $105.58

Chemical Identifiers

Chemical Name or Material alpha-Cyano-4-methoxycinnamic acid
CAS 1519-55-7
Molecular Formula C11H9NO3
Formula Weight 203.2
MDL Number MFCD00051722
Beilstein 2110241
Synonym 4-Methoxybenzylidenecyanoacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0529906

 
alfa aesar™
L0529906
5g Each for $105.58
Description & Specifications

Specifications

Formula Weight 203.2
MDL Number MFCD00051722
Chemical Name or Material alpha-Cyano-4-methoxycinnamic acid
Assay ≥98%
Molecular Formula C11H9NO3
CAS 1519-55-7
Beilstein 2110241
Synonym 4-Methoxybenzylidenecyanoacetic acid
Health Hazard 1 Danger
Melting Point ≈228°C
Quantity 5g
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.