Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Conotoxin SIA

For cellular and molecular biology applications

Specifications

Product Type alpha-Conotoxin SIA
Molecular Weight 1455.66
Storage Requirements 2° to 8°C
Format Lyophilized powder
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66745EXD

 
alfa aesar™
J66745EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66745MCR

 
alfa aesar™
J66745MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type alpha-Conotoxin SIA
Molecular Weight 1455.66
Storage Requirements 2° to 8°C
Format Lyophilized powder
Regulatory Status RUO
Purification Method HPLC