Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Conotoxin MI

For cellular and molecular biology applications

Specifications

Product Type alpha-Conotoxin MI
Molecular Weight 1493.71
Storage Requirements 2° to 8°C
Format Lyophilized powder
Sequence Gly-Arg-Cys-Cys-His-Pro-Ala-Cys-Gly-Lys-Asn-Tyr-Ser-Cys-NH2 (Disulfide bridges: Cys3-Cys8, Cys4-Cys14)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66753EXD

 
alfa aesar™
J66753EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66753MCR

 
alfa aesar™
J66753MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type alpha-Conotoxin MI
Molecular Weight 1493.71
Storage Requirements 2° to 8°C
Format Lyophilized powder
Sequence Gly-Arg-Cys-Cys-His-Pro-Ala-Cys-Gly-Lys-Asn-Tyr-Ser-Cys-NH2 (Disulfide bridges: Cys3-Cys8, Cys4-Cys14)
Regulatory Status RUO
Purification Method HPLC