Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Conotoxin IMI

For cellular and molecular biology applications

Specifications

Product Type alpha-Conotoxin IMI
Molecular Weight 1351.56
Storage Requirements 2° to 8°C
Sequence Gly-Cys-Cys-Ser-Asp-Pro-Arg-Cys-Ala-Trp-Arg-Cys-NH2 (Disulfide bridges: Cys2-Cys8, Cys3-Cys12)
Format Lyophilized powder
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66642EXD

 
alfa aesar™
J66642EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66642MCR

 
alfa aesar™
J66642MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type alpha-Conotoxin IMI
Molecular Weight 1351.56
Storage Requirements 2° to 8°C
Sequence Gly-Cys-Cys-Ser-Asp-Pro-Arg-Cys-Ala-Trp-Arg-Cys-NH2 (Disulfide bridges: Cys2-Cys8, Cys3-Cys12)
Format Lyophilized powder
Regulatory Status RUO
Purification Method HPLC