missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Conotoxin GI

For cellular and molecular biology applications

$157.18 - $236.26

Specifications

Product Type alpha-Conotoxin GI
Molecular Weight 1437.60
Storage Requirements 2° to 8°C
Sequence Glu-Cys-Cys-Asn-Pro-Ala-Cys-Gly-Arg-His-Tyr-Ser-Cys-NH2 (Disulfide bridges: Cys2-Cys7, Cys3-Cys13)
Format Lyophilized powder
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66628EXD
View Documents
Alfa Aesar™
J66628EXD
0.5mg Each for $157.18
 
AAJ66628MCR
View Documents
Alfa Aesar™
J66628MCR
1mg Each for $236.26
 
Specifications

Specifications

alpha-Conotoxin GI
2° to 8°C
Lyophilized powder
HPLC
1437.60
Glu-Cys-Cys-Asn-Pro-Ala-Cys-Gly-Arg-His-Tyr-Ser-Cys-NH2 (Disulfide bridges: Cys2-Cys7, Cys3-Cys13)
RUO

For Research Use Only