Alfa Aesar™ alpha-Conotoxin GI
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66628EXD
 View more versions of this product
Catalog No. AAJ66628EXD
Description
Description
Specifications
Specifications
1437.60
0.5mg
Glu-Cys-Cys-Asn-Pro-Ala-Cys-Gly-Arg-His-Tyr-Ser-Cys-NH2 (Disulfide bridges: Cys2-Cys7, Cys3-Cys13)
2° to 8°C
HPLC
Lyophilized powder
RUO
alpha-Conotoxin GI