Please login to your online account to display your discounted pricing

alpha-Chloro-o-xylene, 99%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC110155000

 View more versions of this product

Catalog No. AC110155000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 140.61
Quantity 500mL
Color Clear colorless to light yellow
Density 1.0800g/mL
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.5405 to 1.5425 (20°C, 589nm)
Chemical Name or Material α-Chloro-o-xylene
Assay Percent Range 99%
Molecular Formula C8H9Cl
MDL Number MFCD00000899
CAS 552-45-4
Synonym 2-Methylbenzyl chloride
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Infrared Spectrum Authentic
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Danger
Linear Formula CH3C6H4CH2Cl
Health Hazard 3 GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material α-Chloro-o-xylene
CAS 552-45-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H9Cl
Linear Formula CH3C6H4CH2Cl
Formula Weight 140.61
MDL Number MFCD00000899
Accessories

GHS Signal Word: Danger

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
May cause respiratory irritation.

SDS & Safety Documents