Please login to your online account to display your discounted pricing

alpha-Chloralose, 98+%, beta anomer ca 15%, Alfa Aesar™

$71.12 - $71.12

Chemical Identifiers

Chemical Name or Material alpha-Chloralose
CAS 15879-93-3
Molecular Formula C8H11Cl3O6
Formula Weight 309.53
MDL Number MFCD00005542
Beilstein 85418
Merck Index 14,2072
Synonym 1,2-O-(2,2,2-Trichloroethylidene)-alpha-D-glucofuranose
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA1149222

 
alfa aesar
A1149222
Each for $71.12
Description & Specifications

Specifications

Formula Weight 309.53
Melting Point 180° to 184°C
Quantity 100g
Chemical Name or Material alpha-Chloralose
TSCA TSCA
Assay ≥98%
Assay Percent Notes beta anomer ≈15%
Molecular Formula C8H11Cl3O6
MDL Number MFCD00005542
CAS 15879-93-3
Beilstein 85418
Merck Index 14,2072
Synonym 1,2-O-(2,2,2-Trichloroethylidene)-alpha-D-glucofuranose
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H332;
Toxic if swallowed.
Harmful if inhaled.
Optical Rotation +19° (c=2 in ethanol)
Health Hazard 3 GHS P Statement
P261-P301+P310-P304+P340-P312-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number FM9450000

Applications
It has been employed widely as an animal anesthetic in the laboratory setting. alpha-chloralose is a true anesthetic or an immobilizing agent with sedative-hypnotic properties has important implications. Canada geese (Branta canadensis) and blue geese (Chen caerulescens) were caught with alpha-chloralose, an oral hypnotic administered on bait. Wild turkeys (Meleagris gallopavo) were captured with alpha-chloralose, an oral anesthetic.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P304+P340-P312-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H332;
Toxic if swallowed.
Harmful if inhaled.