missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

alpha-Bromophenylacetic Acid, TCI America™

$42.78 - $114.58

Chemical Identifiers

CAS 4870-65-9
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00004206
InChI Key WAKFRZBXTKUFIW-UHFFFAOYSA-N
Synonym alpha-bromophenylacetic acid, bromo phenyl acetic acid, .alpha.-bromophenylacetic acid, bromophenylacetic acid, bromo-phenyl-acetic acid, bromo-phenylacetic acid, dl-.alpha.-bromophenylacetic acid, benzeneacetic acid, .alpha.-bromo, phenylbromoacetic acid, bromo phenyl aceticacid
PubChem CID 97919
IUPAC Name 2-bromo-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B09845G View Documents TCI America
B09845G
5g Each for $42.78
 
B098425G View Documents TCI America
B098425G
25g Each for $114.58
 
Specifications

Chemical Identifiers

4870-65-9
215.046
WAKFRZBXTKUFIW-UHFFFAOYSA-N
97919
C1=CC=C(C=C1)C(C(=O)O)Br
C8H7BrO2
MFCD00004206
alpha-bromophenylacetic acid, bromo phenyl acetic acid, .alpha.-bromophenylacetic acid, bromophenylacetic acid, bromo-phenyl-acetic acid, bromo-phenylacetic acid, dl-.alpha.-bromophenylacetic acid, benzeneacetic acid, .alpha.-bromo, phenylbromoacetic acid, bromo phenyl aceticacid
2-bromo-2-phenylacetic acid

Specifications

White-Yellow
5g
C8H7BrO2
3261
WAKFRZBXTKUFIW-UHFFFAOYSA-N
2-bromo-2-phenylacetic acid
97919
Crystal-Powder at 20°C
alpha-Bromophenylacetic Acid
81°C
4870-65-9
MFCD00004206
alpha-bromophenylacetic acid, bromo phenyl acetic acid, .alpha.-bromophenylacetic acid, bromophenylacetic acid, bromo-phenyl-acetic acid, bromo-phenylacetic acid, dl-.alpha.-bromophenylacetic acid, benzeneacetic acid, .alpha.-bromo, phenylbromoacetic acid, bromo phenyl aceticacid
C1=CC=C(C=C1)C(C(=O)O)Br
215.046
215.05
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1715&(9)-1263

TSCA : No