Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Bromocinnamaldehyde, 98%

$141.61 - $141.61

Chemical Identifiers

Chemical Name or Material alpha-Bromocinnamaldehyde
CAS 5443-49-2
Molecular Formula C9H7BrO
Formula Weight 211.06
MDL Number MFCD00006965
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1423022

 
alfa aesar™
A1423022
100g Each for $141.61
Description & Specifications

Specifications

Formula Weight 211.06
Chemical Name or Material alpha-Bromocinnamaldehyde
Assay 98%
Molecular Formula C9H7BrO
MDL Number MFCD00006965
CAS 5443-49-2
Health Hazard 1 Warning
Melting Point 68° to 72°C
Quantity 100g
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number GD6480000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.