missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

alpha-Bromo-gamma-butyrolactone, TCI America™

$48.89 - $447.64

Chemical Identifiers

CAS 5061-21-2
Molecular Formula C4H5BrO2
Molecular Weight (g/mol) 164.986
MDL Number MFCD00005387
InChI Key LFJJGHGXHXXDFT-UHFFFAOYSA-N
Synonym 2-bromo-4-butanolide, alpha-bromo-gamma-butyrolactone, 2-bromobutyrolactone, 3-bromodihydrofuran-2 3h-one, 2 3h-furanone, 3-bromodihydro, 3-bromo-2-furanone, bromobutyrolactone, 2 3h-furanone, bromodihydro, .alpha.-bromo-.gamma.-butyrolactone, .alpha.-bromobutyrolactone
PubChem CID 95463
IUPAC Name 3-bromooxolan-2-one
SMILES C1COC(=O)C1Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B122825G View Documents TCI America
B122825G
25g Each for $48.89
 
B1228500G View Documents TCI America
B1228500G
500g Each for $447.64
 
Specifications

Chemical Identifiers

5061-21-2
164.986
LFJJGHGXHXXDFT-UHFFFAOYSA-N
95463
C1COC(=O)C1Br
C4H5BrO2
MFCD00005387
2-bromo-4-butanolide, alpha-bromo-gamma-butyrolactone, 2-bromobutyrolactone, 3-bromodihydrofuran-2 3h-one, 2 3h-furanone, 3-bromodihydro, 3-bromo-2-furanone, bromobutyrolactone, 2 3h-furanone, bromodihydro, .alpha.-bromo-.gamma.-butyrolactone, .alpha.-bromobutyrolactone
3-bromooxolan-2-one

Specifications

140°C
25g
C4H5BrO2
2-bromo-4-butanolide, alpha-bromo-gamma-butyrolactone, 2-bromobutyrolactone, 3-bromodihydrofuran-2 3h-one, 2 3h-furanone, 3-bromodihydro, 3-bromo-2-furanone, bromobutyrolactone, 2 3h-furanone, bromodihydro, .alpha.-bromo-.gamma.-butyrolactone, .alpha.-bromobutyrolactone
C1COC(=O)C1Br
164.986
164.99
≥95.0% (GC)
Yellow
5061-21-2
MFCD00005387
LFJJGHGXHXXDFT-UHFFFAOYSA-N
3-bromooxolan-2-one
95463
Clear Liquid at 20°C
alpha-Bromo-gamma-butyrolactone
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6756

TSCA : Yes