missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-gamma-butyrolactone 95.0+%, TCI America™

Catalog No. B1228500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1228500G 500 g
B122825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1228500G Supplier TCI America Supplier No. B1228500G
Only null left

Chemical Identifiers

CAS 5061-21-2
Molecular Formula C4H5BrO2
Molecular Weight (g/mol) 164.99
MDL Number MFCD00005387
InChI Key LFJJGHGXHXXDFT-UHFFFAOYNA-N
Synonym 2-bromo-4-butanolide, alpha-bromo-gamma-butyrolactone, 2-bromobutyrolactone, 3-bromodihydrofuran-2 3h-one, 2 3h-furanone, 3-bromodihydro, 3-bromo-2-furanone, bromobutyrolactone, 2 3h-furanone, bromodihydro, .alpha.-bromo-.gamma.-butyrolactone, .alpha.-bromobutyrolactone
PubChem CID 95463
IUPAC Name 3-bromooxolan-2-one
SMILES BrC1CCOC1=O

Specifications

Color Yellow
Boiling Point 140°C
Quantity 500 g
Formula Weight 164.99
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Bromo-gamma-butyrolactone
EINECSNumber (5)-6756
TSCA Yes