missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Bromo-gamma-butyrolactone, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 5061-21-2 |
|---|---|
| Molecular Formula | C4H5BrO2 |
| Molecular Weight (g/mol) | 164.99 |
| MDL Number | MFCD00005387 |
| InChI Key | LFJJGHGXHXXDFT-UHFFFAOYNA-N |
| Synonym | 2-bromo-4-butanolide, alpha-bromo-gamma-butyrolactone, 2-bromobutyrolactone, 3-bromodihydrofuran-2 3h-one, 2 3h-furanone, 3-bromodihydro, 3-bromo-2-furanone, bromobutyrolactone, 2 3h-furanone, bromodihydro, .alpha.-bromo-.gamma.-butyrolactone, .alpha.-bromobutyrolactone |
| PubChem CID | 95463 |
| IUPAC Name | 3-bromooxolan-2-one |
| SMILES | BrC1CCOC1=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAA1734414
|
N/A | 25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AAA1734422
|
N/A | 100g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C4H5BrO2 | |
| MFCD00005387 | |
| 2-bromo-4-butanolide, alpha-bromo-gamma-butyrolactone, 2-bromobutyrolactone, 3-bromodihydrofuran-2 3h-one, 2 3h-furanone, 3-bromodihydro, 3-bromo-2-furanone, bromobutyrolactone, 2 3h-furanone, bromodihydro, .alpha.-bromo-.gamma.-butyrolactone, .alpha.-bromobutyrolactone | |
| 3-bromooxolan-2-one |
Specifications
| 5061-21-2 | |
| 137°C to 139°C (6mmHg) | |
| C4H5BrO2 | |
| MFCD00005387 | |
| Moisture sensitive | |
| LFJJGHGXHXXDFT-UHFFFAOYNA-N | |
| 3-bromooxolan-2-one | |
| 95463 | |
| 97% |
| 1.994 | |
| >110°C (230°F) | |
| 1.508 | |
| 107800 | |
| 2-bromo-4-butanolide, alpha-bromo-gamma-butyrolactone, 2-bromobutyrolactone, 3-bromodihydrofuran-2 3h-one, 2 3h-furanone, 3-bromodihydro, 3-bromo-2-furanone, bromobutyrolactone, 2 3h-furanone, bromodihydro, .alpha.-bromo-.gamma.-butyrolactone, .alpha.-bromobutyrolactone | |
| BrC1CCOC1=O | |
| 164.99 | |
| 164.99 | |
| alpha-Bromo-gamma-butyrolactone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 225-764-4
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only