Learn More
alpha-Bromo-4-fluorophenylacetic acid, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6669206
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
alpha-Bromo-4-fluorophenylacetic acid | |
96°C to 100°C | |
C8H6BrFO2 | |
5 g | |
2-bromo-2-4-fluorophenyl acetic acid, alpha-bromo-4-fluorophenylacetic acid, bromo 4-fluorophenyl acetic acid, alpha-bromo-4-fluorophenyl acetic acid, 2-bromanyl-2-4-fluorophenyl ethanoic acid, alpha-bromo-4-fluorophenylacetic acid 9 | |
C1=CC(=CC=C1C(C(=O)O)Br)F | |
233.036 | |
233.03 | |
Powder |
29270-33-5 | |
White to Brown | |
MFCD08276759 | |
UN3261 | |
ATILMAUMZRFROS-UHFFFAOYSA-N | |
2-bromo-2-(4-fluorophenyl)acetic acid | |
16217752 | |
96% |
Chemical Identifiers
29270-33-5 | |
233.036 | |
ATILMAUMZRFROS-UHFFFAOYSA-N | |
16217752 | |
C1=CC(=CC=C1C(C(=O)O)Br)F |
C8H6BrFO2 | |
MFCD08276759 | |
2-bromo-2-4-fluorophenyl acetic acid, alpha-bromo-4-fluorophenylacetic acid, bromo 4-fluorophenyl acetic acid, alpha-bromo-4-fluorophenyl acetic acid, 2-bromanyl-2-4-fluorophenyl ethanoic acid, alpha-bromo-4-fluorophenylacetic acid 9 | |
2-bromo-2-(4-fluorophenyl)acetic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only