missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Bromo-2-chlorophenylacetic acid, 98%

$15.89 - $15.89

Chemical Identifiers

CAS 29270-30-2
Molecular Formula C8H6BrClO2
Formula Weight 249.49
MDL Number MFCD03428513
UN Number UN3261
Chemical Name or Material alpha-Bromo-2-chlorophenylacetic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2551806 SDS Alfa Aesar™
B2551806
5g Each for $15.89
AAB2551814 SDS Alfa Aesar™
B2551814
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

29270-30-2
249.49
UN3261
C8H6BrClO2
MFCD03428513
alpha-Bromo-2-chlorophenylacetic acid

Specifications

29270-30-2
249.49
UN3261
5g
98%
C8H6BrClO2
MFCD03428513
105° to 108°C
alpha-Bromo-2-chlorophenylacetic acid

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.