Please login to your online account to display your discounted pricing

alpha beta TCR Mouse anti-Human, PerCP-eFluor 710, Clone: IP26, eBioscience™

Mouse Monoclonal Antibody

$166.75 - $427.75

Specifications

Antigen alpha beta TCR
Clone IP26
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123989

 
life technologies
LS46998641
25 Tests Each for $166.75

501123990

 
life technologies
LS46998642
100 Tests Each for $427.75
Description & Specifications

Specifications

Antigen alpha beta TCR
Clone IP26
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The IP26 monoclonal antibody reacts with the alpha beta chain of human TCR.The alpha beta TCR is expressed by the majority of peripheral T cells.