Please login to your online account to display your discounted pricing

alpha beta TCR Mouse anti-Human, PE-Cy7, Clone: IP26, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $390.05

Specifications

Antigen alpha beta TCR
Clone IP26
Host Species Mouse
Gene Alias TCR ab
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123481

 
life technologies
LS25998641
25 tests Each for $158.05

501123482

 
life technologies
LS25998642
100 tests Each for $390.05
Description & Specifications

Specifications

Antigen alpha beta TCR
Clone IP26
Host Species Mouse
Gene Alias TCR ab
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (1μg)/test

The IP26 monoclonal antibody reacts with the human alpha beta TCR dimer. The alpha beta TCR is expressed by the majority of peripheral T cells.