missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

alpha beta TCR Mouse anti-Human, FITC, Clone: IP26, eBioscience™

Mouse Monoclonal Antibody

$104.28 - $240.24

Specifications

Antigen alpha beta TCR
Clone IP26
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122199 Life Technologies
LS11998641
25 Tests Each for $104.28
50245254 Life Technologies
LS11998642
100 Tests Each for $240.24
Specifications

Description

The IP26 monoclonal antibody reacts with the alpha beta chain of human TCR.The alpha beta TCR is expressed by the majority of peripheral T cells.
Description & Specifications

Specifications

alpha beta TCR
Murine
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (1μg)/test
IP26
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer