Please login to your online account to display your discounted pricing

alpha beta TCR Mouse anti-Human, Clone: IP26, eBioscience™

Mouse Monoclonal Antibody

$71.05 - $143.55

Specifications

Antigen alpha beta TCR
Clone IP26
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122899

 
life technologies
LS14998682
100μg Each for $143.55

5013968

 
life technologies
LS14998680
25μg Each for $71.05
Description & Specifications

Specifications

Antigen alpha beta TCR
Clone IP26
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The IP26 monoclonal antibody reacts with the alpha beta chain of human TCR.The alpha beta TCR is expressed by the majority of peripheral T cells.