missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Benzylsuccinic acid, 98+%

$49.97 - $125.80

Chemical Identifiers

CAS 884-33-3
Molecular Formula C11H12O4
Formula Weight 208.22
MDL Number MFCD00055798
Beilstein 1966188
Chemical Name or Material alpha-Benzylsuccinic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1284603 SDS Alfa Aesar™
L1284603
1g Each for $125.80
AAL12846MD SDS Alfa Aesar™
L12846MD
250mg Each for $49.97
Specifications Description & Specifications

Chemical Identifiers

884-33-3
208.22
1966188
C11H12O4
MFCD00055798
alpha-Benzylsuccinic acid

Specifications

884-33-3
208.22
1966188
1g
≥98%
C11H12O4
MFCD00055798
161° to 163°C
alpha-Benzylsuccinic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning