missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Benzylsuccinic acid, 98+%

Manufacturer:  Alfa Aesar™ L12846MD

 View more versions of this product

Catalog No. AAL12846MD



Specifications

Specifications

alpha-Benzylsuccinic acid
250mg
C11H12O4
1966188
GTOFKXZQQDSVFH-UHFFFAOYSA-N
2-benzylbutanedioic acid
3858
208.22
161°C to 163°C
884-33-3
MFCD00055798
2-benzylsuccinic acid, benzylsuccinic acid, dl-benzylsuccinic acid, alpha-benzylsuccinic acid, d,l-benzylsuccinic acid, phenylmethyl butanedioic acid, beta-carboxybenzenebutanoic acid, butanedioic acid,2-phenylmethyl, .alpha.-benzylsuccinic acid, benzenebutanoic acid, .beta.-carboxy
C1=CC=C(C=C1)CC(CC(=O)O)C(=O)O
208.213
CHEBI:16054
≥98%

Chemical Identifiers

884-33-3
208.213
GTOFKXZQQDSVFH-UHFFFAOYSA-N
3858
2-benzylbutanedioic acid
C11H12O4
MFCD00055798
2-benzylsuccinic acid, benzylsuccinic acid, dl-benzylsuccinic acid, alpha-benzylsuccinic acid, d,l-benzylsuccinic acid, phenylmethyl butanedioic acid, beta-carboxybenzenebutanoic acid, butanedioic acid,2-phenylmethyl, .alpha.-benzylsuccinic acid, benzenebutanoic acid, .beta.-carboxy
CHEBI:16054
C1=CC=C(C=C1)CC(CC(=O)O)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents