missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

alpha-Benzoin oxime, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 105530250

 View more versions of this product

Catalog No. AC105530250


 

Specifications Description & Specifications

Specifications

α-Benzoin oxime
97.5
Authentic
Crystalline Powder
98%
C6H5CH(OH)C(=NOH)C6H5
2-Hydroxy-1, 2-diphenylethanone oxime (α-form)
White to almost white
441-38-3
100.0
Glass bottle
≥97.5%
C14H13NO2
227.26
150° to 155°C
25g

Chemical Identifiers

441-38-3
100.0
C14H13NO2
227.26
α-Benzoin oxime
97.5
98%
C6H5CH(OH)C(=NOH)C6H5
2-Hydroxy-1, 2-diphenylethanone oxime (α-form)
≥97.5%