Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Bag Cell Peptide (1-7)

For cellular and molecular biology applications

Specifications

Product Type alpha-Bag Cell Peptide (1-7)
Molecular Weight 922.08
Storage Requirements -30° to -10°C
Sequence Ala-Pro-Arg-Leu-Arg-Phe-Tyr
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66473MA

 
alfa aesar™
J66473MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66473MB

 
alfa aesar™
J66473MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type alpha-Bag Cell Peptide (1-7)
Molecular Weight 922.08
Storage Requirements -30° to -10°C
Sequence Ala-Pro-Arg-Leu-Arg-Phe-Tyr
Format Solid
Regulatory Status RUO
Purification Method HPLC