missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Angelicalactone, 98%

$38.34 - $261.68

Chemical Identifiers

CAS 591-12-8
Molecular Formula C5H6O2
Formula Weight 98.1
MDL Number MFCD00005375
Beilstein 108394
Merck Index 14,647
Synonym 4-Hydroxy-3-pentenoic acid gamma-lactone
Chemical Name or Material alpha-Angelicalactone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0760206 SDS Alfa Aesar™
L0760206
5g Each for $38.34
AAL0760214 SDS Alfa Aesar™
L0760214
25g Each for $102.51
AAL0760222 SDS Alfa Aesar™
L0760222
100g Each for $261.68
Specifications

Description

Applications
alpha-Angelicalactone acts as acancer chemopreventive agent. It is used in the synthesis of glutathione and the activity of glutathione-S-transferase and UDP-glucunonosyltransferase detoxification enzymes in esophagus, stomach, intestine, and liver. It is used in oral care formulations to round out mint notes and in coffee flavors.

Solubility
Soluble in water (5g/100mL) and alcohol.

Notes
Moisture-sensitive. Store under inert gas. Protect from moisture. Incompatible materials are Oxidizing agents.
Description & Specifications

Chemical Identifiers

591-12-8
98.1
108394
4-Hydroxy-3-pentenoic acid gamma-lactone
C5H6O2
MFCD00005375
14,647
alpha-Angelicalactone

Specifications

591-12-8
98.1
108394
14,647
53° to 54°C (10mmHg)
68°C (154°F)
98%
C5H6O2
MFCD00005375
17° to 18°C
4-Hydroxy-3-pentenoic acid gamma-lactone
5g
alpha-Angelicalactone

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : LU5075000

TSCA : TSCA