missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

alpha-Amino-gamma-butyrolactone Hydrobromide 98.0+%, TCI America™

$48.89 - $143.61

Chemical Identifiers

CAS 6305-38-0
Molecular Formula C4H8BrNO2
Molecular Weight (g/mol) 182.017
MDL Number MFCD00012723
InChI Key MKLNTBLOABOJFZ-UHFFFAOYSA-N
Synonym alpha-amino-gamma-butyrolactone hydrobromide, 3-aminooxolan-2-one hydrobromide, hsls hydrobromide, 3-aminodihydrofuran-2 3h-one hydrobromide, 2-amino-4-butyrolactone, 2 3h-furanone, hydrobromide, 2 3h-furanone, 3-aminodihydro-, hydrobromide, 2 3h-furanone,3-aminodihydro-, hydrobromide 1:1, hydrobromide alpha-amino-gamma-butyrolactone, l-homoserine lactone hbr
PubChem CID 73508
IUPAC Name 3-aminooxolan-2-one;hydrobromide
SMILES C1COC(=O)C1N.Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A02831G View Documents TCI America
A02831G
1g Each for $48.89
 
A02835G View Documents TCI America
A02835G
5g Each for $143.61
 
Specifications

Chemical Identifiers

6305-38-0
182.017
MKLNTBLOABOJFZ-UHFFFAOYSA-N
73508
C1COC(=O)C1N.Br
C4H8BrNO2
MFCD00012723
alpha-amino-gamma-butyrolactone hydrobromide, 3-aminooxolan-2-one hydrobromide, hsls hydrobromide, 3-aminodihydrofuran-2 3h-one hydrobromide, 2-amino-4-butyrolactone, 2 3h-furanone, hydrobromide, 2 3h-furanone, 3-aminodihydro-, hydrobromide, 2 3h-furanone,3-aminodihydro-, hydrobromide 1:1, hydrobromide alpha-amino-gamma-butyrolactone, l-homoserine lactone hbr
3-aminooxolan-2-one;hydrobromide

Specifications

White
1g
C4H8BrNO2
alpha-amino-gamma-butyrolactone hydrobromide, 3-aminooxolan-2-one hydrobromide, hsls hydrobromide, 3-aminodihydrofuran-2 3h-one hydrobromide, 2-amino-4-butyrolactone, 2 3h-furanone, hydrobromide, 2 3h-furanone, 3-aminodihydro-, hydrobromide, 2 3h-furanone,3-aminodihydro-, hydrobromide 1:1, hydrobromide alpha-amino-gamma-butyrolactone, l-homoserine lactone hbr
C1COC(=O)C1N.Br
182.017
182.02
≥98.0% (N,T)
237°C
6305-38-0
MFCD00012723
MKLNTBLOABOJFZ-UHFFFAOYSA-N
3-aminooxolan-2-one;hydrobromide
73508
Crystal-Powder at 20°C
alpha-Amino-gamma-butyrolactone Hydrobromide
Safety and Handling

Safety and Handling

TSCA : No