Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha,alpha-Diphenyl-N-methyl-L-prolinol, 97%, ee 99+%

$130.84 - $395.59

Chemical Identifiers

Chemical Name or Material alpha,alpha-Diphenyl-N-methyl-L-prolinol
CAS 110529-22-1
Molecular Formula C18H21NO
Formula Weight 267.37
MDL Number MFCD00145245
Beilstein 4749387
Synonym (S)-alpha,alpha-Diphenyl-1-methylpyrrolidine-2-methanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1939903

  SDS
alfa aesar™
L1939903
1g Each for $395.59

AAL19399MD

  SDS
alfa aesar™
L19399MD
250mg Each for $130.84
Description & Specifications

Specifications

Formula Weight 267.37
Chemical Name or Material alpha,alpha-Diphenyl-N-methyl-L-prolinol
Assay 97%
MDL Number MFCD00145245
CAS 110529-22-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Optical Rotation +52° (c=1 in Chloroform)
Assay Percent Notes ee ≥99%
Molecular Formula C18H21NO
Beilstein 4749387
Synonym (S)-alpha,alpha-Diphenyl-1-methylpyrrolidine-2-methanol
Recommended Storage 2° to 8°C
Melting Point 66° to 69°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C