Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha,alpha-Diphenyl-N-methyl-D-prolinol, 97%, ee 99+%

$128.08 - $387.24

Chemical Identifiers

Chemical Name or Material alpha,alpha-Diphenyl-N-methyl-D-prolinol
CAS 144119-12-0
Molecular Formula C18H21NO
Formula Weight 267.37
MDL Number MFCD00145217
Beilstein 1535407
Synonym (R)-alpha,alpha-Diphenyl-1-methylpyrrolidine-2-methanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL19398MD

  SDS
alfa aesar™
L19398MD
250mg Each for $128.08

AAL1939803

  SDS
alfa aesar™
L1939803
1g Each for $387.24
Description & Specifications

Specifications

Formula Weight 267.37
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
CAS 144119-12-0
Chemical Name or Material alpha,alpha-Diphenyl-N-methyl-D-prolinol
Molecular Formula C18H21NO
Quantity 250mg
Melting Point 68° to 71°C
Recommended Storage 2° to 8°C
Optical Rotation −52° (c=1 in Chloroform)
Beilstein 1535407
MDL Number MFCD00145217
Synonym (R)-alpha,alpha-Diphenyl-1-methylpyrrolidine-2-methanol
Health Hazard 1 Warning
Assay Percent Notes ee ≥99%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C