missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

alpha,alpha-Diphenyl-4-piperidinomethanol hydrochloride, 98%, ACROS Organics™

$75.35 - $222.40

Chemical Identifiers

CAS 1798-50-1
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C18H21NO·HCl
Formula Weight 303.82
MDL Number MFCD00039039
Chemical Name or Material α, α-Diphenyl-4-piperidinomethanol hydrochloride, 98%
Assay ≥97.5% (on dry substance), ≥97.5% (HPLC)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC124981000 SDS Thermo Fisher Scientific
124981000
Glass bottle 100g Each for $222.40
AC124980250 SDS Thermo Fisher Scientific
124980250
Glass bottle 25g Each for $75.35
Specifications Description & Specifications

Chemical Identifiers

1798-50-1
100.0
C18H21NO·HCl
MFCD00039039
≥97.5% (on dry substance), ≥97.5% (HPLC)
97.5
98%
303.82
α, α-Diphenyl-4-piperidinomethanol hydrochloride, 98%

Specifications

303.82
100g
13, 895
97.5
10% max.
1798-50-1
MFCD00039039
α, α-Diphenyl-4-piperidinomethanol hydrochloride, 98%
Crystalline Powder
White to off-white
100.0
Glass bottle
Authentic
C18H21NO·HCl
98%
≥97.5% (on dry substance), ≥97.5% (HPLC)

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 217-284-9

RTECSNumber : TN0925000

TSCA : TSCA