missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

alpha,alpha'-Dibromo-m-xylene, 97%, Acros Organics

Manufacturer:  Thermo Fisher Scientific AC155050250

 View more versions of this product

Catalog No. AC155050250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

α, α'-Dibromo-m-xylene
Glass bottle
White to Brown
25g
96.0
96% min. (HPLC)
C6H4(CH2Br)2
1,3-bis bromomethyl benzene
OXHOPZLBSSTTBU-UHFFFAOYSA-N
1,3-bis(bromomethyl)benzene
69373
Crystalline Powder and Chunks
Authentic
135.0°C to 140.0°C (20.0mmHg)
75.0°C to 77.0°C
626-15-3
100.0
C8H8Br2
05,374
Solubility in water: insoluble
C1=CC(=CC(=C1)CBr)CBr
263.95
263.95
97%

Chemical Identifiers

626-15-3
263.95
1,3-bis bromomethyl benzene
1,3-bis(bromomethyl)benzene
C8H8Br2
OXHOPZLBSSTTBU-UHFFFAOYSA-N
69373
C1=CC(=CC(=C1)CBr)CBr
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Danger

EINECSNumber : 210-931-6

SDS Documents