missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

alpha,alpha′-Bis(4-methylbenzyloxy)-p-xylene, TCI America™

$330.00 - $1,103.08

Chemical Identifiers

CAS 136861-46-6
Molecular Formula C24H26O2
Molecular Weight (g/mol) 346.47
MDL Number MFCD00143577
InChI Key IJMQICMKIDWWAK-UHFFFAOYSA-N
Synonym p-Xylylene Glycol Bis(4-methylbenzyl) Ether
PubChem CID 21659525
IUPAC Name 1,4-bis[(4-methylphenyl)methoxymethyl]benzene
SMILES CC1=CC=C(C=C1)COCC2=CC=C(C=C2)COCC3=CC=C(C=C3)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B14821G View Documents TCI America
B14821G
1g Each for $330.00
 
B14825G View Documents TCI America
B14825G
5g Each for $1,103.08
 
Specifications

Chemical Identifiers

136861-46-6
346.47
IJMQICMKIDWWAK-UHFFFAOYSA-N
21659525
CC1=CC=C(C=C1)COCC2=CC=C(C=C2)COCC3=CC=C(C=C3)C
C24H26O2
MFCD00143577
p-Xylylene Glycol Bis(4-methylbenzyl) Ether
1,4-bis[(4-methylphenyl)methoxymethyl]benzene

Specifications

White
1g
C24H26O2
p-Xylylene Glycol Bis(4-methylbenzyl) Ether
CC1=CC=C(C=C1)COCC2=CC=C(C=C2)COCC3=CC=C(C=C3)C
346.47
346.47
≥98.0% (GC)
100°C
136861-46-6
MFCD00143577
IJMQICMKIDWWAK-UHFFFAOYSA-N
1,4-bis[(4-methylphenyl)methoxymethyl]benzene
21659525
Crystal-Powder at 20°C
alpha,alpha′-Bis(4-methylbenzyloxy)-p-xylene
Safety and Handling

Safety and Handling

TSCA : No