missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

alpha,alpha′-Bis(4-hydroxyphenyl)-1,4-diisopropylbenzene, TCI America™

$67.22 - $481.27

Chemical Identifiers

CAS 2167-51-3
Molecular Formula C24H26O2
Molecular Weight (g/mol) 346.47
MDL Number MFCD00191315
InChI Key GIXXQTYGFOHYPT-UHFFFAOYSA-N
Synonym 1,4-Bis[2-(4-hydroxyphenyl)-2-propyl]benzene, 4,4′-(1,4-Phenylenediisopropylidene)bisphenol, Bisphenol P
PubChem CID 630355
IUPAC Name 4-[2-[4-[2-(4-hydroxyphenyl)propan-2-yl]phenyl]propan-2-yl]phenol
SMILES CC(C)(C1=CC=C(C=C1)C(C)(C)C2=CC=C(C=C2)O)C3=CC=C(C=C3)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B156325G View Documents TCI America
B156325G
25g Each for $67.22
 
B1563500G View Documents TCI America
B1563500G
500g Each for $481.27
 
Specifications

Chemical Identifiers

2167-51-3
346.47
GIXXQTYGFOHYPT-UHFFFAOYSA-N
630355
CC(C)(C1=CC=C(C=C1)C(C)(C)C2=CC=C(C=C2)O)C3=CC=C(C=C3)O
C24H26O2
MFCD00191315
1,4-Bis[2-(4-hydroxyphenyl)-2-propyl]benzene, 4,4′-(1,4-Phenylenediisopropylidene)bisphenol, Bisphenol P
4-[2-[4-[2-(4-hydroxyphenyl)propan-2-yl]phenyl]propan-2-yl]phenol

Specifications

White-Yellow
25g
C24H26O2
1,4-Bis[2-(4-hydroxyphenyl)-2-propyl]benzene, 4,4′C-(1,4-Phenylenediisopropylidene)bisphenol, Bisphenol P
CC(C)(C1=CC=C(C=C1)C(C)(C)C2=CC=C(C=C2)O)C3=CC=C(C=C3)O
346.47
346.47
≥98.0% (GC)
195°C
2167-51-3
MFCD00191315
GIXXQTYGFOHYPT-UHFFFAOYSA-N
4-[2-[4-[2-(4-hydroxyphenyl)propan-2-yl]phenyl]propan-2-yl]phenol
630355
Crystal-Powder at 20°C
alpha,alpha′-Bis(4-hydroxyphenyl)-1,4-diisopropylbenzene
Safety and Handling

Safety and Handling

EINECSNumber : (4)-1538

TSCA : No