missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

alpha,alpha′-Bis(4-aminophenyl)-1,4-diisopropylbenzene, TCI America™

$77.92 - $672.23

Chemical Identifiers

CAS 2716-10-1
Molecular Formula C24H28N2
Molecular Weight (g/mol) 344.50
MDL Number MFCD00191313
InChI Key HESXPOICBNWMPI-UHFFFAOYSA-N
Synonym 1,4-Bis[2-(4-aminophenyl)-2-propyl]benzene, 4,4′-(1,4-Phenylenediisopropylidene)bisaniline, Bisaniline P
PubChem CID 75930
IUPAC Name 4-[2-[4-[2-(4-aminophenyl)propan-2-yl]phenyl]propan-2-yl]aniline
SMILES CC(C)(C1=CC=C(C=C1)C(C)(C)C2=CC=C(C=C2)N)C3=CC=C(C=C3)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B1562500G View Documents TCI America
B1562500G
500g Each for $672.23
 
B156225G View Documents TCI America
B156225G
25g Each for $77.92
 
Specifications

Chemical Identifiers

2716-10-1
344.50
HESXPOICBNWMPI-UHFFFAOYSA-N
75930
CC(C)(C1=CC=C(C=C1)C(C)(C)C2=CC=C(C=C2)N)C3=CC=C(C=C3)N
C24H28N2
MFCD00191313
1,4-Bis[2-(4-aminophenyl)-2-propyl]benzene, 4,4′-(1,4-Phenylenediisopropylidene)bisaniline, Bisaniline P
4-[2-[4-[2-(4-aminophenyl)propan-2-yl]phenyl]propan-2-yl]aniline

Specifications

Undesignated
500g
C24H28N2
1,4-Bis[2-(4-aminophenyl)-2-propyl]benzene, 4,4′-(1,4-Phenylenediisopropylidene)bisaniline, Bisaniline P
CC(C)(C1=CC=C(C=C1)C(C)(C)C2=CC=C(C=C2)N)C3=CC=C(C=C3)N
344.50
344.50
≥99.0% (GC)
165°C
2716-10-1
MFCD00191313
HESXPOICBNWMPI-UHFFFAOYSA-N
4-[2-[4-[2-(4-aminophenyl)propan-2-yl]phenyl]propan-2-yl]aniline
75930
Crystal-Powder at 20°C
alpha,alpha′-Bis(4-aminophenyl)-1,4-diisopropylbenzene
Safety and Handling

Safety and Handling

EINECSNumber : (4)-1573

RTECSNumber : CY1202650

TSCA : Yes