Please login to your online account to display your discounted pricing

alpha,alpha,alpha-Trifluoro-o-tolyl isocyanate, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  170340250

 View more versions of this product

Catalog No. AC170340250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4700 to 1.4760 (20°C, 589nm)
Density 1.2300g/mL
Color Clear colorless to light yellow
Quantity 25g
Chemical Name or Material α, α, α-Trifluoro-o-tolyl isocyanate
Name Note 97%
CAS 140957
CAS Min % 96.0
CAS Max % 100.0
Physical Form Clear colorless to light yellow liquid
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C8H4F3NO
Linear Formula CF3C6H4NCO
Formula Weight 187.12g/mol
MDL Number MFCD00002008
Synonym 2-Trifluoromethylphenylisocyanate
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Fatal if inhaled.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number NR0201000
EINECS Number 218-925-5
TSCA TSCA

Specifications

Chemical Name or Material α, α, α-Trifluoro-o-tolyl isocyanate
Name Note 97%
CAS 140957
CAS Min % 96.0
CAS Max % 100.0
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C8H4F3NO
Linear Formula CF3C6H4NCO
Formula Weight 187.12g/mol
MDL Number MFCD00002008
Synonym 2-Trifluoromethylphenylisocyanate
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Fatal if inhaled.
Causes skin irritation.
Flammable liquid and vapor.

SDS & Safety Documents