Please login to your online account to display your discounted pricing

alpha,alpha,alpha-Trifluoro-o-tolyl isocyanate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material α, α, α-Trifluoro-o-tolyl isocyanate
Name Note 97%
CAS 140957
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H4F3NO
Linear Formula CF3C6H4NCO
Formula Weight 187.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC170340050

  SDS
thermo fisher scientific
AC170340050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥96.0%
Formula Weight 187.12
Quantity 5g
Color Clear colorless to light yellow
EINECS Number 218-925-5
TSCA TSCA
RTECS Number NR0201000
Density 1.2300g/mL
CAS Min % 96.0
CAS Max % 100.0
CAS 140957
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4700 to 1.4760 (20°C, 589nm)
Assay Percent Range 97%
Molecular Formula C8H4F3NO
Linear Formula CF3C6H4NCO
MDL Number MFCD00002008
Infrared Spectrum Authentic
Synonym 2-Trifluoromethylphenylisocyanate
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Fatal if inhaled.
Causes skin irritation.
Flammable liquid and vapor.
Chemical Name or Material α, α, α-Trifluoro-o-tolyl isocyanate
Name Note 97%