Please login to your online account to display your discounted pricing

alpha,alpha,alpha-Trifluoro-m-tolyl isocyanate, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC170350250

 View more versions of this product

Catalog No. AC170350250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 187.12
Physical Form Liquid
Quantity 25g
Color Clear colorless
Density 1.3360g/mL
CAS Max % 100.0
Refractive Index 1.4670 to 1.4690 (20°C, 589nm)
Linear Formula CF3C6H4NCO
Health Hazard 1 Danger
Molecular Formula C8H4F3NO
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautio
MDL Number MFCD00002020
CAS 329-01-1
Health Hazard 2 GHS P Statement Toxic in contact with skin. Fatal if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes serious eye damage. May cause respiratory irritation. Causes skin irritat
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym 3-(Trifluoromethyl)phenyl isocyanate
Chemical Name or Material α, α, α-Trifluoro-m-tolyl isocyanate
Assay Percent Range 98%

Specifications

Chemical Name or Material α, α, α-Trifluoro-m-tolyl isocyanate
CAS 329-01-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H4F3NO
Linear Formula CF3C6H4NCO
Formula Weight 187.12
MDL Number MFCD00002020
Synonym 3-(Trifluoromethyl)phenyl isocyanate
Accessories

Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautio

GHS P Statement Toxic in contact with skin. Fatal if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes serious eye damage. May cause respiratory irritation. Causes skin irritat

SDS & Safety Documents