missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

alpha,alpha,alpha-Trifluoro-m-tolyl isocyanate, 98%, ACROS Organics™

Chemical Identifiers

CAS 329-01-1
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C8H4F3NO
Linear Formula CF3C6H4NCO
Formula Weight 187.12
MDL Number MFCD00002020
Synonym 3-(Trifluoromethyl)phenyl isocyanate
Chemical Name or Material α, α, α-Trifluoro-m-tolyl isocyanate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC170350250 SDS Thermo Fisher Scientific
AC170350250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

329-01-1
100.0
C8H4F3NO
187.12
3-(Trifluoromethyl)phenyl isocyanate
≥97.5%
97.5
98%
CF3C6H4NCO
MFCD00002020
α, α, α-Trifluoro-m-tolyl isocyanate

Specifications

1.3360g/mL
25g
Authentic
100.0
1.4670 to 1.4690 (20°C, 589nm)
98%
CF3C6H4NCO
MFCD00002020
α, α, α-Trifluoro-m-tolyl isocyanate
Clear colorless
329-01-1
97.5
Glass bottle
Liquid
C8H4F3NO
187.12
3-(Trifluoromethyl)phenyl isocyanate
≥97.5%

Safety and Handling

GHS P Statement Toxic in contact with skin. Fatal if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes serious eye damage. May cause respiratory irritation. Causes skin irritat

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautio

Danger