Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha,alpha,alpha-Tribromoquinaldine, 97%

$57.57 - $803.11

Chemical Identifiers

Chemical Name or Material alpha,alpha,alpha-Tribromoquinaldine
CAS 613-53-6
Molecular Formula C10H6Br3N
Formula Weight 379.89
MDL Number MFCD00006755
Beilstein 147638
Synonym 2-(Tribromomethyl)quinoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2521809

  SDS
alfa aesar™
B2521809
10g Each for $57.57

AAB2521818

  SDS
alfa aesar™
B2521818
50g Each for $183.82

AAB2521830

  SDS
alfa aesar™
B2521830
250g Each for $803.11
Description & Specifications

Specifications

Formula Weight 379.89
Chemical Name or Material alpha,alpha,alpha-Tribromoquinaldine
TSCA TSCA
Assay 97%
Molecular Formula C10H6Br3N
MDL Number MFCD00006755
CAS 613-53-6
Beilstein 147638
Synonym 2-(Tribromomethyl)quinoline
Health Hazard 1 Warning
Melting Point 128° to 131°C
Quantity 10g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number VC3250750
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.