Learn More
alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 137970250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
α,α,α',α'-Tetrabromo-o-xylene | |
13209-15-9 | |
100.0 | |
96% min. (GC) | |
C8H6Br4 | |
MFCD00000131 | |
05,367 | |
Solubility in water: insoluble | |
C1=CC=C(C(=C1)C(Br)Br)C(Br)Br | |
421.75 | |
421.75 |
97% | |
96.0 | |
Authentic | |
Plastic bottle | |
C6H4(CHBr2)2 | |
25g | |
1,2-bis dibromomethyl benzene, alpha,alpha,alpha',alpha'-tetrabromo-o-xylene, benzene, 1,2-bis dibromomethyl, unii-3wg2h68vx4, o-xylylene tetrabromide, alpha,alpha,alpha,alpha-tetrabromo-o-xylene, a,a,a',a'-tetrabromo-o-xylene, o-xylene, alpha,alpha,alpha',alpha'-tetrabromo, alpha,alpha,beta,beta-tetrabromo-o-xylene, acmc-209boi | |
LNAOKZKISWEZNY-UHFFFAOYSA-N | |
1,2-bis(dibromomethyl)benzene | |
83234 | |
97% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 236-176-2
TSCA : TSCA
RUO â Research Use Only